Skip to Content
Merck
All Photos(2)

Documents

P9789

Sigma-Aldrich

1,2-Dipalmitoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt

≥99% (TLC)

Synonym(s):

1,2-Dihexadecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, L-α-Phosphatidyl-DL-glycerol, dipalmitoyl, DPPG-Na, Hexadecanoic acid 1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, PG(16:0/16:0)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C38H74O10PNa
CAS Number:
Molecular Weight:
744.95
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25

biological source

semisynthetic

Quality Level

Assay

≥99% (TLC)

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C38H75O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-37(41)45-33-36(34-47-49(43,44)46-32-35(40)31-39)48-38(42)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h35-36,39-40H,3-34H2,1-2H3,(H,43,44);/q;+1/p-1/t35?,36-;/m1./s1

InChI key

LDWIWSHBGAIIMV-ODZMYOIVSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Forms classical lipid bilayers. At low temperature, however, an unusual and much thinner bilayer forms where the hydrocarbon chains are interdigitated.

Other Notes

PG(16:0/16:0)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service