Skip to Content
Merck
All Photos(1)

Key Documents

N0912

Sigma-Aldrich

NAEPA

≥98 (TLC), solid

Synonym(s):

(Z)-N-[2-(Phosphonooxy)ethyl]-9-octadecenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40NO5P
CAS Number:
Molecular Weight:
405.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

form

solid

color

white

mp

75.8-84.0 °C

solubility

DMSO: soluble 28 mg/mL
H2O: soluble

storage temp.

2-8°C

SMILES string

CCCCCCCC\C=C\CCCCCCCC(=O)NCCOP(O)(O)=O

InChI

1S/C20H40NO5P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(22)21-18-19-26-27(23,24)25/h9-10H,2-8,11-19H2,1H3,(H,21,22)(H2,23,24,25)/b10-9+

InChI key

BCSUWOZFWWBYSX-MDZDMXLPSA-N

Biochem/physiol Actions

Selective lysophosphatidic acid-1 (LPA1) receptor agonist; LPA mimetic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David B Banks et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 38(5), 857-868 (2017-04-28)
The blood-brain barrier is a microvascular network that (1) provides neuroprotection from metabolic and environmental toxins and (2) limits the delivery of therapeutics to the central nervous system (CNS). The ATP-binding cassette transporter P-glycoprotein contributes to the latter by actively
Izabela Woclawek-Potocka et al.
Experimental biology and medicine (Maywood, N.J.), 234(8), 986-993 (2009-06-06)
Lysophosphatidic acid (LPA) has been shown to be a potent modulator of prostaglandin (PG) secretion during the luteal phase of the estrous cycle in the bovine endometrium in vivo. The aims of the present study were to determine the cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service