MTCP80511S
MTC™ Bio Divided Reservoir
capacity 50 mL, sterile, pack of 100 ea (individually wrapped)
Synonym(s):
reagent dish, reagent holder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121522
NACRES:
NB.12
Recommended Products
sterility
sterile
packaging
pack of 100 ea (individually wrapped)
manufacturer/tradename
MTC™ Bio P8051-1S
capacity
50 mL
Looking for similar products? Visit Product Comparison Guide
General description
Reservoir, divided, 50ml sterile , Individually wrapped, 100/cs
Features and Benefits
- Graduated inner walls
- Micro-trough U-shaped bottoms
- Pouring spouts on all 4 corners
Legal Information
MTC is a trademark of MTC Bio
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service