Skip to Content
Merck
All Photos(1)

Key Documents

MTCH6505

MTC Bio Halo pipette tips

capacity 5 mL

Synonym(s):

pipetting tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
capacity:
5 mL
sterility:
sterile
feature:
DNase free
RNase free
packaging:
pack of 100 ea
manufacturer/tradename:
MTC Bio H6505

material

clear
polypropylene

sterility

sterile

feature

DNase free
RNase free

packaging

pack of 100 ea

manufacturer/tradename

MTC Bio H6505

capacity

5 mL

Looking for similar products? Visit Product Comparison Guide

General description

5mL Pipette Tips for Halo Pipettors, 100/pk
MTC Bio’s Macro 5 and 10mL tips are specially designed for an ideal and comfortable fit on Macro size pipettes. Supplied in bags, these tips are made of ultra-clear polypropylene and are guaranteed to create the most ideal seal on ProPette™ Pipettors.

Features and Benefits

  • 10-40% lighter than leading competitors
  • 4 Digit micrometer with DriftGuard protection
  • Exclusive 1250µL model for extended length tips
  • User-enabled calibration port
  • Compatible with universal tips

Legal Information

Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service