Skip to Content
Merck
All Photos(2)

Key Documents

J0250

Sigma-Aldrich

Jack beans

Whole Beans

Synonym(s):

Canavalia Gladiata, Canavalia Seeds, Jack Bean Seeds

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.81

Quality Level

form

solid

Looking for similar products? Visit Product Comparison Guide

General description

Jack beans or Carnavalia ensiformis is a leguminous plant rich in proteins that produces mass amounts of green fodder. It is a source of feed supplement for dwarf goats.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agronomic and nutritive evaluation of jack beans (Canavalia ensiformis) for fodder in the derived savannah zone of Nigeria
Olaniran O OjebryiandO A
International Journal of Agriculture and Biology, 2(12), 1059-1063 (2007)
Effect of spontaneous fermentation on the chemical composition of thermally treated Jack Beans (Canavalia ensiformis L.)
Gabriel, RAO and Akharaiyi F C
International journal of biological and chemical sciences, 1(2), 91-97 (2007)
Christina J Welch et al.
Current protocols in protein science, 101(1), e113-e113 (2020-09-04)
Glycosylated proteins, namely glycoproteins and proteoglycans (collectively called glycoconjugates), are indispensable in a variety of biological processes. The functions of many glycoconjugates are regulated by their interactions with another group of proteins known as lectins. In order to understand the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service