Skip to Content
Merck
All Photos(4)

Key Documents

HPA023430

Sigma-Aldrich

Anti-CEP55 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C10orf3, Anti-CT111, Anti-Centrosomal protein 55kDa, Anti-FLJ10540

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
WB
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

PLVTFQGETENREKVAASPKSPTAALNESLVECPKCNIQYPATEHRDLLVHVEYCS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEP55(55165)

General description

The gene CEP55 (Centrosomal protein of 55kDa), also referred to as C10orf3, encodes a 464 amino acid containing protein with highest expression in testis and thymus. This coiled-coil protein has a molar mass of 55kDa. The gene is mapped to human chromosome 10q23.33.

Immunogen

centrosomal protein 55kDa recombinant protein epitope signature tag (PrEST)

Application

Anti-CEP55 antibody produced in rabbit has been used in immunohistochemistry.
All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene CEP55 (Centrosomal protein of 55kDa) encodes a protein that is found to be localized to the mother centriole during interphase. It plays a crucial role in microtubule nucleation, mitotic exit and cytokinesis. The protein CEP55 also elicits the cell transformation by stimulating the phosphatidylinositol 3′-kinase (PI3K)/AKT pathway. It participates in the regulation of cell-cycle and is found in certain tumor tissues, such as hepatocellular carcinoma and gastric cancer. Additionally, it is also involved in midbody structure during cytokinesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70314

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linbang Wang et al.
Frontiers in oncology, 10, 554138-554138 (2021-03-16)
Cancer stem cells (CSCs) represent a subset of tumor cells that are responsible for recurrence and metastasis of tumors. These cells are resistant to radiotherapy and chemotherapy. Immunotherapeutic strategies that target CSCs specifically have provided initial results; however, the mechanism
CEP55 contributes to human gastric carcinoma by regulating cell proliferation.
Tao J
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 35, 4389-4399 (2014)
C-H Chen et al.
Oncogene, 26(29), 4272-4283 (2007-01-24)
A significant challenge in the post-genomic era is how to prioritize differentially expressed and uncharacterized novel genes found in hepatocellular carcinoma (HCC) microarray profiling. One such category is cell cycle regulated genes that have only evolved in higher organisms but
Megan Fabbro et al.
Developmental cell, 9(4), 477-488 (2005-10-04)
Centrosomes in mammalian cells have recently been implicated in cytokinesis; however, their role in this process is poorly defined. Here, we describe a human coiled-coil protein, Cep55 (centrosome protein 55 kDa), that localizes to the mother centriole during interphase. Despite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service