Skip to Content
Merck
All Photos(2)

Key Documents

HPA023319

Sigma-Aldrich

Anti-CNTROB antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Centrobin, Anti-Centrosomal BRCA2-interacting protein, Anti-LYST-interacting protein 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

LEELFPRYTSLRPGPPLNPPDFQGLRDALDSEHTRRKHCERHIQSLQTRVLELQQQLAVAVAADRKKDTMIEQLDKTLARVVEGWNRHEAERTEVLRGLQEEHQAAELTRSKQQE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CNTROB(116840)

General description

The gene CNTROB (centrobin) is mapped to human chromosome 17p13. The protein localizes in the centrioles and is also present in the cytoplasm.

Immunogen

Centrobin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CNTROB (centrobin) is involved in the centriole assembly, bipolar mitotic spindle assembly, microtubule polymerization and asymmetric cell division. It associates with CEP152 (centrosomal protein 152) and CPAP (centrosomal P4.1-associated protein). The centrobin-CPAP interaction is needed for the centriole duplication. Mutations in CNTROB are associated with breast cancer susceptibility.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75804

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Gottardo et al.
Journal of cell science, 129(14), 2726-2731 (2016-05-29)
The uncoordinated (unc) gene product, a potential ortholog of mammalian orofaciodigital syndrome 1 (Ofd1), is involved in the assembly of the ciliary axoneme in Drosophila and it is, therefore, constrained to cell types that have ciliary structures, namely type 1
Andrea Zatkova et al.
Genes, chromosomes & cancer, 48(6), 510-520 (2009-03-24)
AML/MDS patients carrying 11q amplifications involving the mixed lineage leukemia gene (MLL) locus are characterized by a complex aberrant karyotype (CAK) frequently including deletions within 5q, 17p, and 7q, older age and fast progression of the disease with extremely poor
Radhika Gudi et al.
The Journal of biological chemistry, 289(22), 15166-15178 (2014-04-05)
Centriole duplication is the process by which two new daughter centrioles are generated from the proximal end of preexisting mother centrioles. Accurate centriole duplication is important for many cellular and physiological events, including cell division and ciliogenesis. Centrosomal protein 4.1-associated
Wonjung Shin et al.
Cell cycle (Georgetown, Tex.), 14(12), 1925-1931 (2015-06-18)
Centrobin resides in daughter centriole and play a critical role in centriole duplication. Nucleation and stabilization of microtubules are known biological activities of centrobin. Here, we report a specific localization of centrobin outside the centrosome. Centrobin was associated with the
Hui Wang et al.
Breast cancer research and treatment, 136(1), 241-251 (2012-09-25)
Centrosome aberrations have been suggested to cause chromosomal instability and aneuploidy, and eventually promote cancer development. The Centrobin and Nek2 proteins interact with each other and both are involved in centrosome duplication and chromosome segregation. This study aimed to investigate whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service