Skip to Content
Merck
All Photos(2)

Key Documents

HPA012393

Sigma-Aldrich

Anti-ADGRA2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DKFZp434C211, Anti-DKFZp434J0911, Anti-FLJ14390, Anti-GPR124, Anti-KIAA1531, Anti-TEM5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SPTDSYLGSSRNSPGAGLQLEGEPMLTPSEGSDTSAAPLSEAGRAGQRRSASRDSLKGGGALEKESHRRSYPLNAASLNGAPKGGKYDDVTLMGAEVASGGCMKTGLWK

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR124(25960)

Immunogen

adhesion G protein-coupled receptor A2

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GPR124 (G protein-coupled receptor 124) is an adhesion G-protein-coupled receptor protein belonging to the adhesion family of G-protein-coupled receptors. It consists of a RGD (Arg-Gly-Asp) motif in its extracellular end. It is expressed in the surface of endothelial cells and pericytes at the time of angiogenesis. It has been reported that thrombin cleaves the protein into a 60kDa fragment, which binds to the RGD-dependent integrins. This interaction may associate with various cellular functions such as adhesion and migration during angiogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71166

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Vallon et al.
The Biochemical journal, 441(3), 937-944 (2011-10-22)
TEM5 (tumour endothelial marker 5; also known as GPR124) is an adhesion G-protein-coupled receptor containing a cryptic RGD motif in its extracellular domain. TEM5 is expressed in endothelial cells and pericytes during angiogenesis. In the present paper, we report that
Mario Vallon et al.
The Journal of biological chemistry, 281(45), 34179-34188 (2006-09-20)
Tumor endothelial marker (TEM) 5 is a member of the adhesion family of G-protein-coupled receptors and up-regulated in endothelial cells during tumor and physiologic angiogenesis. Here, we report that TEM5 is expressed on the surface of endothelial cells. A soluble
Ioannis A Voutsadakis
Journal of clinical medicine, 9(10) (2020-09-30)
Amplification of the locus 8p11.23 has been observed in cancer and genes of this locus, including ZNF703 (Zinc finger protein 703), NSD3 (Nuclear receptor binding SET domain protein 3) and FGFR1 (Fibroblast growth factor receptor 1), have been put forward
Characteristics and Prognosis of 8p11.23-Amplified Squamous Lung Carcinomas.
Voutsadakis
Journal of clinical medicine, 12 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service