Skip to Content
Merck
All Photos(2)

Key Documents

HPA011935

Sigma-Aldrich

Anti-SLC2A8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GLUT-8, Anti-Glucose transporter type 8, Anti-Glucose transporter type X1, Anti-Solute carrier family 2, facilitated glucose transporter member 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
3
technique(s):
immunohistochemistry: 1:20- 1:50

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

PRFLLTQHRRQEAMAALRFLWGSEQGWEDPPIGAEQSFHLALLRQPGIYKP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC2A8(29988)

Immunogen

Solute carrier family 2, facilitated glucose transporter member 8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC2A8 (Solute carrier family 2, member 8) is a cell-surface localized, class III glucose and fructose transporter. It is involved in the glucose transport activity, hepatocyte fructose transport, and fructose-induced macrosteatosis. It is highly expressed in testis, brain, liver, heart, and other oxidative tissues. Its expression is also associated with the endosomes, lysosomes, and membranes of the endoplasmic reticulum. It facilitates the transport of glucose and its derivatives through intracellular membranes. It also mediates the translocation of fructose to its primary metabolism site i. e. hepatocytes. In testis, it provides fuel to mature spermatozoa for the sperm cell functioning. Alerations in SLC2A8 leads to non-alcoholic fatty liver disease (NAFLD).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71275

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian J Debosch et al.
The Journal of biological chemistry, 289(16), 10989-10998 (2014-02-13)
Non-alcoholic fatty liver disease (NAFLD) is the most common liver disease in the world, and it is thought to be the hepatic manifestation of the metabolic syndrome. Excess dietary fructose causes both metabolic syndrome and NAFLD in rodents and humans
Annette Schürmann et al.
Cell and tissue research, 307(2), 237-242 (2002-02-15)
The glucose transporter 8 (GLUT8) is a recently identified member of the family of sugar transport facilitators. In human tissues GLUT8 is predominantly expressed in testis in a gonadotropin-dependent manner. It is shown here that the onset of mRNA synthesis
Stefan Schmidt et al.
American journal of physiology. Endocrinology and metabolism, 296(4), E614-E618 (2009-01-30)
GLUT8 is a class III sugar transporter predominantly expressed in testis and brain. In contrast to the class I and class II transporters, hydrophobicity plots predict a short extracellular loop between transmembrane domain (TM)1 and TM2 and a long extracellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service