Skip to Content
Merck
All Photos(10)

Key Documents

HPA004747

Sigma-Aldrich

Anti-FLNB antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

ABP-278, FLN1L, Fh1, Filamin-B, beta, LRS1, TABP, TAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FLNB(2317)

General description

FLNB (filamin B, β) is a 280kDa cytoskeletal protein consists of the NH2-terminal actin-binding domain, a backbone of 24 tandem repeats, and two "hinge" regions. It belongs to the filamin family.

Immunogen

Filamin-B recombinant protein epitope signature tag (PrEST)

Sequence
KEPGEYAVHIMCDDEDIKDSPYMAFIHPATGGYNPDLVRAYGPGLEKSGCIVNNLAEFTVDPKDAGKAPLKIFAQDGEGQRIDIQMKNRMDGTYACSYTPVKAIKHTIAVVWG

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FLNB (filamin B, β) is involved in the organization of actin fibers and endothelial cell motility. It interacts with the cytoplasmic tail of glycoprotein (Gp) Ibα in endothelial cells and platelets. It helps to arrange various transmembrane proteins to the actin cytoskeleton for the scaffold like structure.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70089

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Takafuta et al.
The Journal of biological chemistry, 273(28), 17531-17538 (1998-07-04)
We have cloned and sequenced a 9.4-kilobase cDNA specifying a new 280-kDa protein interacting with the cytoplasmic tail of glycoprotein (Gp) Ibalpha and showing considerable homology to actin-binding protein 280 (ABP-280) and chicken retinal filamin. We term this protein human
Marie-France Nissou et al.
Journal of neuro-oncology, 113(2), 239-249 (2013-04-02)
Most of our knowledge regarding glioma cell biology comes from cell culture experiments. For many years the standards for glioma cell culture were the use of cell lines cultured in the presence of serum and 20 % O2. However, in
A van der Flier et al.
Biochimica et biophysica acta, 1538(2-3), 99-117 (2001-05-05)
Filamins are a family of high molecular mass cytoskeletal proteins that organize filamentous actin in networks and stress fibers. Over the past few years it has become clear that filamins anchor various transmembrane proteins to the actin cytoskeleton and provide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service