Skip to Content
Merck
All Photos(9)

Key Documents

HPA003326

Sigma-Aldrich

Anti-CD2AP antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Adapter protein CMS antibody produced in rabbit, Anti-CD2-associated protein antibody produced in rabbit, Anti-Cas ligand with multiple SH3 domains antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VDFDSLTVRTSKETDVVNFDDIASSENLLHLTANRPKMPGRRLPGRFNGGHSPTHSPEKILKLPKEEDSANLKPSELKKDTCYSPKPSVYLSTPSSASKANTTAFLTPLEIKAKVETDDVKKNSLDELRAQIIELLCIVEALKKDHGKEL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD2AP(23607)

Looking for similar products? Visit Product Comparison Guide

Immunogen

CD2-associated protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CD2AP (CD2-associated protein) gene encodes a protein that forms a component of the filtration complex in the kidney and its haploinsufficiency serves as a determinant of human susceptibility to glomerular disease. The protein is an adaptor molecule that mediates T cell receptor signaling and podocyte homeostasis. It inhibits TGF-β-induced apoptosis in podocytes. It may function as a direct adaptor between the actin cytoskeleton and cell membrane proteins, such as ephrin and polycystin-2, and regulate the actin cytoskeleton.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85224

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piyush Gondaliya et al.
Journal of diabetes and metabolic disorders, 19(1), 13-27 (2020-06-19)
As epigenetic modifications like chromatin histone modifications have been suggested to play a role in the pathophysiology of Diabetic Nephropathy (DN) and are also found to be regulated by microRNAs. Our main purpose was to explore the role of microRNA
Jeong M Kim et al.
Science (New York, N.Y.), 300(5623), 1298-1300 (2003-05-24)
Loss of CD2-associated protein (CD2AP), a component of the filtration complex in the kidney, causes death in mice at 6 weeks of age. Mice with CD2AP haploinsufficiency developed glomerular changes at 9 months of age and had increased susceptibility to
Sanna Lehtonen et al.
American journal of physiology. Renal physiology, 283(4), F734-F743 (2002-09-10)
CD2-associated protein (CD2AP) is an adapter protein associating with several membrane proteins, including nephrin, mutated in congenital nephrotic syndrome of the Finnish type, and polycystin-2, mutated in type 2 autosomal dominant polycystic kidney disease. Both proteins have critical roles in
Mario Schiffer et al.
The Journal of biological chemistry, 279(35), 37004-37012 (2004-06-24)
CD2-associated protein (CD2AP) is an adaptor molecule involved in T cell receptor signaling and podocyte homeostasis. CD2AP-deficient mice develop nephrotic syndrome and renal failure caused by glomerulosclerosis. Here we report that increased transforming growth factor-beta1 (TGF-beta1) expression and apoptosis were
J Nicholas Cochran et al.
Human molecular genetics, 24(23), 6667-6674 (2015-09-12)
CD2-associated protein (CD2AP) is a leading genetic risk factor for Alzheimer's disease, but little is known about the function of CD2AP in the brain. We studied CD2AP(-/-) mice to address this question. Because CD2AP(-/-) mice normally die by 6 weeks

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service