Skip to Content
Merck
All Photos(1)

Documents

H7250

Sigma-Aldrich

Histamine dihydrochloride

≥99% (TLC), powder, endogenous histamine receptor agonist

Synonym(s):

2-(4-Imidazolyl)ethylamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3 · 2HCl
CAS Number:
Molecular Weight:
184.07
Beilstein:
3624116
EC Number:
MDL number:
UNSPSC Code:
12352116
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.77

product name

Histamine dihydrochloride, ≥99% (TLC), powder

Assay

≥99% (TLC)

form

powder

color

white to off-white

mp

249-252 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].NCCc1c[nH]cn1

InChI

1S/C5H9N3.2ClH/c6-2-1-5-3-7-4-8-5;;/h3-4H,1-2,6H2,(H,7,8);2*1H

InChI key

PPZMYIBUHIPZOS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Histamine dihydrochloride is a derivative of histamine. It prevents the formation of ROS (reactive oxygen species), which inhibits the activation of T cells and natural killer cells. Histamine dihydrochloride is implicated in tumor cell apoptosis. It prevents the relapse of acute myeloid leukemia in patients.

Application

Histamine dihydrochloride has been used:
  • as a orthograde cotransmitter in producing excitatory postsynaptic potential (EPSP)
  • to determine its level in fish using HPLC (high performance liquid chromatography)
  • to assess the tracheal response to antigens in guinea-pigs

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunotherapy with histamine dihydrochloride for the prevention of relapse in acute myeloid leukemia
Martner A, et al.
Expert Review of Hematology, 3(4), 381-391 (2010)
Determination of some quality criteria of cold storaged marinated anchovy under vacuum and modified atmosphere conditions
Guncsen U, et al.
Turkish Journal of Fisheries and Aquatic Sciences, 11(2), 282-291 (2011)
Maternal dietary antigens and the immune response in the offspring of the guinea-pig.
Telemo E, et al.
Immunology, 62(1), 35-35 (1987)
Nitric oxide as an orthograde cotransmitter at central synapses of Aplysia: responses of isolated neurons in culture
Jacklet JW and Koh HY
American Zoologist, 41(2), 282-291 (2001)
Histamine H4 receptor: a Novel Drug Target For Immunoregulation and Inflammation (2013)

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service