GE28-9232-44
MidiSpin Adapter
Cytiva 28-9232-44, pack of 10 pieces
All Photos(1)
About This Item
Recommended Products
feature
wetted part: no
packaging
pack of 10 pieces
manufacturer/tradename
Cytiva 28-9232-44
General description
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
Adapters are polypropylene rings designed for use with the gravity columns PD MiniTrap G-25, PD MidiTrap G-25, and PD-10 Desalting Columns when the spin protocol is used. MiniSpin Adapter fits PD MiniTrap G-25 columns, MidiSpin Adapter fits PD MidiTrap G-25 columns, and PD-10 Spin Adapter fits PD-10 Desalting Columns.
Adapters are polypropylene rings designed for use with the gravity columns PD MiniTrap G-25, PD MidiTrap G-25, and PD-10 Desalting Columns when the spin protocol is used. MiniSpin Adapter fits PD MiniTrap G-25 columns, MidiSpin Adapter fits PD MidiTrap G-25 columns, and PD-10 Spin Adapter fits PD-10 Desalting Columns.
Features and Benefits
- Convenient adapters, enable spinning of all gravity columns.
Storage Class Code
13 - Non Combustible Solids
Protocols
This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.