Skip to Content
Merck
All Photos(3)

Documents

G4625

Sigma-Aldrich

Calcium D-gluconate monohydrate

≥98% (titration)

Synonym(s):

D-Gluconic acid calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22CaO14 · H2O
CAS Number:
Molecular Weight:
448.39
Beilstein:
3919646
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic (organic)

Quality Level

Assay

≥98% (titration)

form

powder

technique(s)

HPLC: suitable

color

white to faint yellow

solubility

water: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[Ca]OC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/2C6H12O7.Ca.H2O/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;;/h2*2-5,7-11H,1H2,(H,12,13);;1H2/q;;+2;/p-2/t2*2-,3-,4+,5-;;/m11../s1

InChI key

XLNFVCRGJZBQGX-XRDLMGPZSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • High-Specific Surface Area Microspheres: Calcium D-gluconate monohydrate is used to synthesize yolk-shell structured microspheres of calcium phosphate, demonstrating its utility in creating materials with high-specific surface areas for biomedical applications, such as adsorption processes in cell culture systems (Cao et al., 2020).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service