Skip to Content
Merck
All Photos(1)

Key Documents

EHU011031

Sigma-Aldrich

MISSION® esiRNA

targeting human TLR7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGAAGTTGGCTTCTGCTCAAATGCCAGAACTTCTGTAGAAAGTTATGAACCCCAGGTCCTGGAACAATTACATTATTTCAGATATGATAAGTATGCAAGGAGTTGCAGATTCAAAAACAAAGAGGCTTCTTTCATGTCTGTTAATGAAAGCTGCTACAAGTATGGGCAGACCTTGGATCTAAGTAAAAATAGTATATTTTTTGTCAAGTCCTCTGATTTTCAGCATCTTTCTTTCCTCAAATGCCTGAATCTGTCAGGAAATCTCATTAGCCAAACTCTTAATGGCAGTGAATTCCAACCTTTAGCAGAGCTGAGATATTTGGACTTCTCCAACAACCGGCTTGATTTACTCCATTCAACAGCATTTGAAGAGCTTCACAAACTGGAAGTTCTGGATATAAGCAGTAATAGCCATTATTTTCAATCAGAAGGAATTACTCATATGCTAAACTTTACCAAGAACCTAAAGGTTCTGCAGAAACTGATGATGAACGACAATGACATCTCTTCCTCCACCAGCAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuoyan Zheng et al.
JCI insight, 5(14) (2020-07-24)
TLR7 has been linked to the pathogenesis of glomerulonephritis, but its precise roles are not clear. In this study, we evaluated the roles of TLR7 in IgA nephropathy (IgAN). TLR7 proteins were abundant in CD19+ B cells infiltrated in the
Yayi Huang et al.
Revista da Associacao Medica Brasileira (1992), 65(8), 1067-1073 (2019-09-19)
Diabetes is a risk factor for acute kidney injury (AKI). However, its mechanism of pathogenesis has not been elucidated. The aim of the study was to investigate the role of inflammation and the toll-like receptor 7 (TLR7) in ischemic AKI
Federica Liotti et al.
Cancers, 13(4) (2021-02-14)
Pattern recognition receptors (PRR) promote inflammation but also its resolution. We demonstrated that a specific PRR-formyl peptide receptor 1 (FPR1)-sustains an inflammation resolution response with anti-angiogenic and antitumor potential in gastric cancer. Since toll-like receptor 7 (TLR7) is crucial in
Yun-Ru Liao et al.
Biochemical and biophysical research communications, 493(2), 1136-1142 (2017-08-28)
Diabetic retinopathy (DR) is a major microvascular complication of diabetes, resulting in neuronal dysfunction, retinal vascular leakage, and apoptosis within the retina. Innate immunity plays an important role in the pathogenesis of type 2 diabetes (T2D) and related complications. The
Ryoichiro Nishibayashi et al.
PloS one, 10(6), e0129806-e0129806 (2015-06-18)
Interleukin-12 (IL-12) is an important cytokine for the immunomodulatory effects of lactic acid bacteria (LAB). Using murine immune cells, we previously reported that the RNA of Enterococcus faecalis EC-12, a LAB strain exerting probiotic-like beneficial effects, is the major IL-12-inducing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service