Skip to Content
Merck
All Photos(1)

Key Documents

CRISPR15H

Sigma-Aldrich

Lenti CRISPR Non-Target gRNA Control Transduction Particles, High Titer (LV02 vector)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

form

liquid

Quality Level

packaging

vial of 8 x 25 μL

concentration

(5x108 TU/ml (via p24 titering assay))

application(s)

CRISPR

shipped in

dry ice

storage temp.

−70°C

Related Categories

General description

The Lenti CRISPR Non-Target gRNA Control Transduction Particles (LV02 vector) include a gRNA sequence that does not target known human, mouse and rat genes. It is useful as a negative control in experiments using CRISPR lentiviral clones. The lenti CRISPR non-target control uses a dual-component system consisting of U6-driven guide RNA (non-target) and Ef1a-driven Puromycin resistance cassette. This control requires Cas9 be delivered by a separate vector.

Lentiviral-based particles permit efficient infection and integration of the construct into differentiated and non-dividing cells, such as neurons and dendritic cells, overcoming low transfection and integration difficulties when using these cell lines. Self-inactivating replication incompetent viral particles are produced in packaging cells (HEK293T) by co-transfection with compatible packaging and envelope plasmids.

Application

Functional Genomics/Screening/Target Validation

Other Notes

Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells, respectively.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service