CLS4217
Corning® Step-R Repeating Pipettor
non-sterile
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.02
Recommended Products
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Corning 4217
Related Categories
General description
The Corning Step-R Repeating Pipettor is suitable to dispense volatile, dense, viscous, or foaming liquids, and for all applications where long dosing series are required. It works with 8 syringe tips sizes to dispense doses from 1 μL up to 5 mL.
- Easy volume setting: Volume selection chart conveniently placed on the handle
- Fast and precise multiple dispensing: Up to 48 pipetting doses without refilling the syringe tip
- Maintenance-free
- Meets ISO 8655 requirements.
- Delivered with QC certificate and 25 and 50 mL syringe tips adapters
Legal Information
Corning is a registered trademark of Corning, Inc.