Skip to Content
Merck
All Photos(1)

Documents

M. dunni (Clone III8C)

94101211, mouse tail, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

M. dunni (Clone III8C), 94101211

biological source

mouse tail

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Not specified

receptors

Ecotropic Murine leukemia virus (MuLV)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse tail, normal fibroblast

Cell Line Description

Derived from the tail tissue of a normal female mouse, Mus dunni. The cells were cloned at passage 14 using Tererasiki plates. M. dunni (Clone III8C) supports the replication of all four classes of murine leukaemia viruses (MuLV); ecotropic, amphotropic, xenotropic and mink cell focus-forming viruses (MCF), showing cytopathic effect. The cells lack most endogenous murine leukaemia virus-related sequences. They are resistant to infection by Moloney MuLV, due to post-translational modifications of the ecotropic MuLV receptor. Confluent cultures will remain more firmly attached in RPMI 1640 or DMEM + 10% FBS during virus assays.

Application

Detection of replication-competent retroviruses in gene therapy.

Culture Medium

McCoy′s 5a + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:8 i.e. seeding at 1-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service