Skip to Content
Merck
All Photos(1)

Key Documents

45.6.TG1.7

90110506, mouse blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

45.6.TG1.7, 90110506

biological source

mouse blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Immunoglobulin G (IgG2b) ( kappa)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c myeloma

Cell Line Description

Derived from a female BALB/c myeloma tumour MPC-11. The parental cell line has mutated during growth in 6-thioguanine to establish the drug resistant line 45.6.TG1.7. This line is HPRT negative and produces mouse IgG2b kappa immunoglobulins.

Application

Fusion partner, immunoglobulin production

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 4-5x100,000 cells/ml; 5% CO2; 37°C. Cells achieve a maximum cell density of 1-1.5x1,000,000 cells/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selection of the right fusion partner cell line is critical to obtain desired attributes of the hybridoma. Explore hybridoma fusion partners cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service