Skip to Content
Merck
All Photos(1)

Key Documents

ATC-10 (Aedes aegypti)

87091801, mosquito larvae, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

ATC-10 (Aedes aegypti), 87091801

biological source

mosquito larvae

growth mode

Adherent aggregates

karyotype

2n = 6

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mosquito larvae

Cell Line Description

Derived from pooled newly hatched Aedes aegypti larvae. The cells are susceptible to a range of arboviruses.

Application

Virus studies: Arboviruses

Culture Medium

Mitsuhashi and Maramorosch Insect Medium (pH 7.0) + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5 x10,000 cells/cm2 using 0.25% trypsin for 5 mins only 28°C. After quenching the trypsin with fresh mediumdislodge the cells by gentle scraping.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service