Recommended Products
Product Name
WEHI 3B, 86013003
biological source
mouse blood
growth mode
Suspension
karyotype
Not specified
morphology
Not specified
products
Interleukin 3 (IL-3)
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse BALB/c myelomonocyte
Cell Line Description
Derived from a BALB/c mouse which has undergone mineral oil injections intended to induce plasma cell tumour development. The original tumour WEHI-3b showed many characteristics of human myelomonocytic leukaemia. The cells produce high levels of IL-3.
Application
Tumourigenicity studies
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Maintain cultures between 1-5x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service