Skip to Content
Merck
All Photos(1)

Key Documents

VCaP Cell Line human

6020201, human prostate (metastasis), Epithelial

Synonym(s):

Human VCaP Cells, VCaP Cell Culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

VCaP Cell Line human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 6020201, human prostate (metastasis), Epithelial, Prostate Cancer Metastasis, 06020201

biological source

human prostate (metastasis)

packaging

tube of 5 μg 06020201-DNA-5UG
pkg of vial of cells 06020201-1VL

growth mode

Adherent

karyotype

Refer to reference (Korenchuk et al., 2001)

morphology

Epithelial

products

Not specified

receptors

Androgen receptor

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Prostate Cancer Metastasis

Cell Line Description

This Vertebral-Cancer of the Prostate (VCaP) cell line was established from prostate cancer tissue harvested from a metastatic lesion to a lumbar vertebral body of a patient with hormone refractory prostate cancer. The cells are reported to express large quantities of prostate specific antigen (PSA). This cell line also expresses prostatic acid phosphatase (PAP), cytokeratin-18 and the androgen receptor, and is androgen sensitive in vitro and in vivo. This cell line offers a cell-based model system of human prostate cancer. The cells are tumourigenic in SCID mice.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 11,12
D16S539: 9
D5S818: 12
D7S820: 9,12
THO1: 9.3
TPOX: 8,11
vWA: 18,19

Culture Medium

DMEM:Ham′s F12 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. Pellet cells by centrifugation and remove trypsin after inactivation of trypsin at each subculture.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service