Skip to Content
Merck
All Photos(1)

Key Documents

C265

Sigma-Aldrich

Monoclonal Anti-CaM Kinase IIα (CaMKIIα) antibody produced in mouse

clone 6G9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CAMKA, Anti-CaMKIINalpha, Anti-CaMKIIalpha, Anti-MRD53, Anti-MRT63

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G9, monoclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

rat, mouse

technique(s)

indirect immunofluorescence: suitable using rat hippocampus
western blot: 0.5-1.0 μg/mL

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

CaM Kinase II α is a Ca2+/calmodulin-dependent kinase that belongs to the serine/threonine protein kinases family. It facilitates the regulation of NMDAR-dependent AMPA receptor trafficking to the synapses. Mouse anti-CaM kinase II α antibody reacts specifically with phosphorylated and non-phosphorylated forms of native and recombinant CaM (Ca2+/calmodulin-dependent) kinase II α subunit in rats. The product has shown no reactivity for phosphorylated or non-phosphorylated β subunit (60kD).

Immunogen

partially purified rat CaM kinase II.

Application

Monoclonal Anti-CaM Kinase IIα (CaMKIIα) antibody produced in mouse has been used in immunohistochemistry and western blotting.

Physical form

Solution in phosphate buffered saline containing 0.05% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ke-Hui Hu et al.
Behavioural brain research, 359, 81-88 (2018-10-27)
Ischemic stroke is a major cause of disability and mortality worldwide, while no unequivocally efficacious drug is currently available to treat post-stroke functional impairments. Animal and clinical investigations suggest that the motor cortex stimulation constitutes a particularly promising approach for
Metabotropic signal transduction for bradykinin in submucosal neurons of guinea pig small intestine
Hu HZ, et al.
Journal of Pharmacology and Experimental Therapeutics, 309(1), 310-319 (2004)
Naohiro Kon et al.
Genes & development, 28(10), 1101-1110 (2014-05-17)
Daily behavioral rhythms in mammals are governed by the central circadian clock, located in the suprachiasmatic nucleus (SCN). The behavioral rhythms persist even in constant darkness, with a stable activity time due to coupling between two oscillators that determine the
Wen Fong Ooi et al.
PLoS genetics, 9(9), e1003795-e1003795 (2013-09-27)
Burkholderia pseudomallei (Bp), the causative agent of the often-deadly infectious disease melioidosis, contains one of the largest prokaryotic genomes sequenced to date, at 7.2 Mb with two large circular chromosomes (1 and 2). To comprehensively delineate the Bp transcriptome, we
Jay F Muller et al.
The Journal of comparative neurology, 519(4), 790-805 (2011-01-20)
The basolateral nucleus of the amygdala receives an extremely dense cholinergic innervation from the basal forebrain that is critical for memory consolidation. Although previous electron microscopic studies determined some of the postsynaptic targets of cholinergic afferents, the majority of postsynaptic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service