C-23260
Skeletal Muscle Cell Growth Medium
Basal Medium, 500 ml
Synonym(s):
Recommended with: C-39365 or C-39360 (for growth medium) or C-39366 (for Differentiation medium).
About This Item
Recommended Products
form
aqueous solution
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
storage temp.
2-8°C
General description
To form a complete Skeletal Muscle Cell Growth Medium, it should be combined with the media supplements: C-39365 or C-39360. To form a complete Skeletal Muscle Differentiation Medium , it should be combined with the media supplements: C-39366
Application
Components
Quality
Storage and Stability
Subculture Routine
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Primary human skeletal muscle cells and culture medium optimized for the in vitro cultivation and differentiation of primary human skeletal muscle cells. Protocols for handling, cell passaging, media, and product use.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service