Key Documents
BR782911
BRAND® Insert strips 9mm for 24 x 6 well BRANDplates®
Inlet Opening System, 4 Inserts/Strip, PET membrane
About This Item
UNSPSC Code:
41122107
NACRES:
NB.22
material
PET membrane
description
cellGrade(TM) plus
sterility
sterile
packaging
pack of 12 ea (strips of 4 inserts, induvidullay packed)
manufacturer/tradename
BRAND 782911
size
6 well
pore size
0.4 μm
binding type
non-treated surface
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)