AXYP96450VY
V-bottom assay plate
size 96 wells, V-bottom yellow polypropylene wells, SBS FootPrint, yellow, maximum volume 500 μL, pkg of 10x5plates/cs
Synonym(s):
96 multiwell plate
About This Item
yellow plate
Recommended Products
material
V-bottom yellow polypropylene wells
yellow plate
sterility
non-sterile
manufacturer/tradename
Axygen Inc P-96-450V-Y
Corning
packaging
pkg of 10x5plates/cs
maximum volume
500 μL
size
96 wells
working volume
450 μL
color
yellow
suitability
suitable for PCR
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service