Skip to Content
Merck
All Photos(1)

Key Documents

AV48717

Sigma-Aldrich

Anti-LIAS antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HUSSY-01, Anti-LAS, Anti-LIP1, Anti-Lipoic acid synthetase, Anti-MGC23245

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
yeast, guinea pig, horse, rat, rabbit, human, dog, mouse, bovine
citations:
1
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

yeast, guinea pig, horse, rat, rabbit, human, dog, mouse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LIAS(11019)

General description

Lipoic acid synthetase (LIAS) a mitochondrial protein that catalyzes the synthesis of α-(+)-lipoic acid synthesis. Deficiency of LIAS is linked to glycine elevation, defects in mitochondrial energy metabolism and neonatal-onset epilepsy.
Rabbit Anti-LIAS antibody recognizes human, mouse, and rat LIAS antibody.

Immunogen

Synthetic peptide directed towards the C terminal region of human LIAS

Application

Rabbit Anti-LIAS antibody is suitable for western blot applications at a concentration of 0.25 μg/ml.

Biochem/physiol Actions

LIAS belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis.The protein encoded by this gene belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.

Sequence

Synthetic peptide located within the following region: EYITPEKFKYWEKVGNELGFHYTASGPLVRSSYKAGEFFLKNLVAKRKTK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes A Mayr et al.
American journal of human genetics, 89(6), 792-797 (2011-12-14)
Lipoic acid is an essential prosthetic group of four mitochondrial enzymes involved in the oxidative decarboxylation of pyruvate, α-ketoglutarate, and branched chain amino acids and in the glycine cleavage. Lipoic acid is synthesized stepwise within mitochondria through a process that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service