Skip to Content
Merck
All Photos(3)

Key Documents

AV41516

Sigma-Aldrich

Anti-SLC22A1 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-1-Oct, Anti-HOCT1, Anti-Oct1_cds, Anti-Solute carrier family 22 (organic cation transporter), member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

61 kDa

species reactivity

bovine, rat, human, horse, guinea pig, rabbit, dog, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC22A1(6580)

General description

Solute carrier family 22 (organic cation transporter), member 1 (SLC22A1, 1-Oct, HOCT1, Oct1-cds) is a transporter of organic cations (OCT) that in addition to endogenous cations include various external origin cationic toxins and drugs. Examples of drugs transported by OCT1 include metformin, amantadine, pramipexole, and, possibly, levodopa.

Specificity

Anti-SLC22A1 (AB1) polyclonal antibody reacts with chicken, pig, bovine, rabbit, human, mouse, rat, and canine solute carrier family 22 (organic cation transporter), member 1 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC22A1

Application

Anti-SLC22A1 (AB1) polyclonal antibody is used to tag solute carrier family 22 (organic cation transporter), member 1 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of solute carrier family 22 (organic cation transporter), member 1 in the transport of small organic cations including a variety of important drugs.

Biochem/physiol Actions

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. SLC22A1 contains twelve putative transmembrane domains and is a plasma integral membrane protein.Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. Two transcript variants encoding two different isoforms have been found for this gene, but only the longer variant encodes a functional transporter.

Sequence

Synthetic peptide located within the following region: LSCVDPLASLATNRSHLPLGPCQDGWVYDTPGSSIVTEFNLVCADSWKLD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianxiang Kevin Han et al.
Molecular pharmacology, 84(2), 182-189 (2013-05-18)
Organic cation transporters (OCTs) are members of the solute carrier 22 family of transporter proteins that are involved in absorption, distribution, and excretion of organic cations. OCT3 is localized in the apical (AP) membrane of enterocytes, but the literature is
Metee Jinakote et al.
Fundamental & clinical pharmacology, 37(4), 833-842 (2023-02-28)
Buspirone, a cationic drug, is an anxiolytic and antidepressant drug. However, whether buspirone and its metabolites are interacted with organic cationic transporter remains uncertain. In this study, we examined the interaction of buspirone and its major metabolites 1-(2-pyrimidinyl)piperazine (1-PP) and
Raymond K Hau et al.
Drug metabolism and disposition: the biological fate of chemicals, 50(6), 770-780 (2022-03-22)
The blood-testis barrier (BTB) is formed by basal tight junctions between adjacent Sertoli cells (SCs) of the seminiferous tubules and acts as a physical barrier to protect developing germ cells in the adluminal compartment from reproductive toxicants. Xenobiotics, including antivirals
Ruijin Shao et al.
Journal of experimental & clinical cancer research : CR, 33, 41-41 (2014-06-03)
Although a number of in vitro studies have demonstrated the antiproliferative, anti-invasive, and antimetastatic effects of metformin in multiple cancer cell types, its cellular and molecular mechanisms of anti-cancer action in the endometrium of women with polycystic ovary syndrome (PCOS)
Xin Li et al.
American journal of translational research, 7(3), 574-586 (2015-06-06)
Conflicting results have been reported regarding whether or not insulin-regulated glucose transporter 4 (GLUT4) is expressed in human and rodent endometria. There is an inverse relationship between androgen levels and insulin-dependent glucose metabolism in women. Hyperandrogenemia, hyperinsulinemia, and insulin resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service