Skip to Content
Merck
All Photos(1)

Key Documents

A5660

Sigma-Aldrich

Aminoallyl-UTP sodium salt

≥75%, lyophilized powder

Synonym(s):

5-[3-Aminoallyl]uridine 5′-triphosphate, AA-UTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N3Na3O15P3
CAS Number:
Molecular Weight:
605.17
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥75%

form

lyophilized powder

SMILES string

[Na+].[Na+].[Na+].NC\C=C\C1=CN([C@@H]2O[C@H](OP([O-])(=O)CP([O-])(=O)CP(O)([O-])=O)[C@@H](O)[C@H]2O)C(=O)NC1=O

InChI

1S/C13H22N3O13P3.3Na/c14-3-1-2-7-4-16(13(20)15-10(7)19)11-8(17)9(18)12(28-11)29-32(26,27)6-30(21,22)5-31(23,24)25;;;/h1-2,4,8-9,11-12,17-18H,3,5-6,14H2,(H,21,22)(H,26,27)(H,15,19,20)(H2,23,24,25);;;/q;3*+1/p-3/b2-1+;;;/t8-,9+,11-,12-;;;/m1.../s1

InChI key

MFRKSTHYXMPZPJ-ZPVWDQBGSA-K

Application

Aminoallyl-UTP sodium salt has been used for the production of labelled cDNA and cRNA for fluorescence detection in microarray.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G J Thompson et al.
Insect molecular biology, 17(6), 657-665 (2009-01-10)
A defining characteristic of eusocial animals is their division of labour into reproductive and nonreproductive specialists. Here, we used a microarray study to identify genes associated with functional sterility in the worker honey bee Apis mellifera. We contrasted gene expression
Genome-wide analysis of genes related to ovary activation in worker honey bees.
Thompson GJ, et al.
Insect Molecular Biology, 17, 657-665 (2008)
The construction and use of bacterial DNA microarrays based on an optimized two-stage PCR strategy.
Postier BL, et al.
BMC Genomics, 4, 23-23 (2003)
Distinctive expression of chemokines and transforming growth factor-beta signaling in human arterial endothelium during atherosclerosis.
Volger OL, et al.
The American Journal of Pathology, 171, 326-337 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service