Skip to Content
Merck
All Photos(1)

Key Documents

3021

Sigma-Aldrich

Transcreener® GDP TR-FRET Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122100
NACRES:
NA.84

shipped in

dry ice

storage temp.

−20°C

General description

The Transcreener® GDP TR-FRET Red Assay is a competitive immunoassay for GDP with a far-red, time-resolved Förster-resonance-energy-transfer (TR-FRET) readout. Because it is highly selective for GDP, the assay can be used with any enzyme that converts GTP to GDP, regardless of what other substrates are used. Examples of enzymes include GTPases and fucosyltransferases. The Transcreener® assay is designed specifically for high-throughput screening (HTS), with a single-addition, mix-and-read format. It offers reagent stability and compatibility with commonly used multimode plate readers. The generic nature of the Transcreener® HTS assay platform eliminates delays involved in assay development for new HTS targets and greatly simplifies compound and inhibitor profiling across multiple target families.

The Transcreener® GDP TR-FRET Red Assay provides the following benefits:
  • Accommodates GTP concentrations ranging from 0.1 μM to 1,000 μM.
  • Excellent data quality (Z′ = 0.7) at low substrate conversion (typically 10–30%).
  • Overcomes the need for time-consuming, one-off assay development for individual members within a group transfer enzyme family by using a single set of assay reagents that detect an invariant product.
  • Time-gated detection method largely eliminates interference that can result from prompt fluorescence of test compounds.
  • Far-red tracer further minimizes interference from fluorescent compounds and light scattering.

View full Transcreener® product list

Quantity

3021-1K = 1,000 assay, 384-well
3021-10K = 10,000 assay, 384-well

Physical form

Kit with buffered aqueous solutions

Legal Information

Transcreener is a registered trademark of BellBrook Labs

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Levek-Motola et al.
Human reproduction (Oxford, England), 20(12), 3469-3475 (2005-08-27)
The pathway of spermatogenesis involves the conversion of diploid stem cells (spermatogonia) to tetraploid primary spermatocytes, followed by meiosis and two cell divisions, first forming diploid secondary spermatocytes and then haploid round spermatids. Differentiation of round spermatids results in spermatozoa
A Balduini et al.
Journal of thrombosis and haemostasis : JTH, 4(4), 848-860 (2006-04-26)
Megakaryopoiesis represents a multi-step, often unclear, process leading to commitment, differentiation, and maturation of megakaryocytes (MKs) that release platelets. To identify the novel genes that might help to clarify the molecular mechanisms of megakaryocytopoiesis and be regarded as potential candidates
Carolin Lepa et al.
Journal of the American Society of Nephrology : JASN, 32(2), 357-374 (2021-01-01)
Injury to kidney podocytes often results in chronic glomerular disease and consecutive nephron malfunction. For most glomerular diseases, targeted therapies are lacking. Thus, it is important to identify novel signaling pathways contributing to glomerular disease. Neurotrophic tyrosine kinase receptor 3
Lauriane Simon et al.
Nucleic acids research, 46(6), 3019-3033 (2018-03-09)
Organized in tandem repeat arrays in most eukaryotes and transcribed by RNA polymerase III, expression of 5S rRNA genes is under epigenetic control. To unveil mechanisms of transcriptional regulation, we obtained here in depth sequence information on 5S rRNA genes
Junji Hamuro et al.
Investigative ophthalmology & visual science, 61(5), 47-47 (2020-05-27)
To clarify the expression profiles of ion channels and transporters of metabolic substrates among heterogeneous cultured human corneal endothelial cells (cHCECs) distinct in their effectiveness in reconstituting the corneal endothelium. Integrated proteomics for cell lysates by liquid chromatography-tandem mass spectrometry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service