Skip to Content
Merck
All Photos(1)

Key Documents

96482

Sigma-Aldrich

Zinc nitrate hexahydrate

purum p.a., crystallized, ≥99.0% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
297.49
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
crystals or chunks
solubility:
water: soluble(lit.)

grade

purum p.a.

Assay

≥99.0% (KT)

form

crystals or chunks

quality

crystallized

solubility

water: soluble(lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg

SMILES string

O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.6H2O.Zn/c2*2-1(3)4;;;;;;;/h;;6*1H2;/q2*-1;;;;;;;+2

InChI key

JGPSMWXKRPZZRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zinc nitrate hexahydrate is an inorganic zinc salt. It is used as a zinc precursor in organic synthesis.

Application

Zinc nitrate hexahydrate is used as a zinc precursor in the preparation of:
  • ZnO (zinc oxide) nanoparticles.
  • Zinc oxide nanorods (ZnO NRs), which is used as a catalyst in the photocatalytic degradation of HPAM (hydrolyzed polyacrylamide).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural and DC electrical investigations of ZnO thin films prepared by spray pyrolysis technique.
Riad AS, et al.
Physica B: Condensed Matter (Amsterdam, Netherlands), 296(4), 319-325 (2001)
Xuemin Qian et al.
Nanoscale research letters, 3(8), 303-307 (2008-01-01)
ZnO nanorod arrays are prepared on a silicon wafer through a multi-step hydrothermal process. The aspect ratios and densities of the ZnO nanorod arrays are controlled by adjusting the reaction times and concentrations of solution. The investigation of field emission
Milna GWA.
Gardner's Commercially Important Chemicals: Synonyms, Trade Names, and Properties, 4153-4153 (2015)
Toward wearable and stretchable fabric-based supercapacitors: novel ZnO and SnO2 nanowires-carbon fibre and carbon paper hybrid structure.
Bae J, et al.
Journal of Solid State Electrochemistry, 19(1), 211-219 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service