Skip to Content
Merck
All Photos(1)

Key Documents

24004

Sigma-Aldrich

Diethyl ether

≥99.5% (GC), puriss, contains ~5 mg/L 2, 6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.07
technique(s):
GC/GC: suitable
bp:
34.6 °C (lit.)

Product Name

Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)

vapor density

2.6 (vs air)

Quality Level

grade

puriss.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

320 °F

contains

~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer

expl. lim.

36.5 %

technique(s)

GC/GC: suitable

impurities

acidic reac. Impurities, in accordance
aldehydes, in accordance
residual solvents, in accordance
≤0.0002% peroxides (as H2O2)
≤0.002% free acid (as CH3COOH)
≤0.002% non-volatile matter
≤0.2% water (Karl Fischer)

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethyl ether may be employed as a solvent for the conversion of secondary alcohols into ketones.

Other Notes

The article number 24004-4X2.5L-R will be discontinued. Please order the single bottle 24004-2.5L-R which is physically identical with the same exact specifications.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diels-Alder reactions in ionic liquids. A safe recyclable alternative to lithium perchlorate-diethyl ether mixtures.
Earle MJ, et al.
Green Chemistry, 1(1), 23-25 (1999)
Diethyl ether (DEE) as a renewable diesel fuel.
Bailey B, et al.
No. 972978. SAE technical paper (1997)
Oxidation of secondary alcohols in diethyl ether with aqueous chromic acid. Convenient procedure for the preparation of ketones in high epimeric purity.
Brown HC, et al.
The Journal of Organic Chemistry, 36(3), 387-390 (1971)
K H Young et al.
Journal of clinical microbiology, 10(6), 852-853 (1979-12-01)
Ethyl acetate appears to be a satisfactory subsitute solvent for diethyl ether in the Formalin-ether sedimentation technique. In comparative studies, concentration of organisms with ethyl acetate was equal to or greater than that with diethyl ether. No distortion or alteration
Eagleson M.
Concise Encyclopedia Chemistry, 471-471 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service