Skip to Content
Merck
All Photos(1)

Key Documents

Y0000454

Diacetylfluorescein

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Fluorescein diacetate, 3,6-Diacetoxyfluoran, Di-O-acetylfluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16O7
CAS Number:
Molecular Weight:
416.38
Beilstein:
62575
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diacetylfluorescein

manufacturer/tradename

EDQM

mp

200-203 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc2c(Oc3cc(OC(C)=O)ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C24H16O7/c1-13(25)28-15-7-9-19-21(11-15)30-22-12-16(29-14(2)26)8-10-20(22)24(19)18-6-4-3-5-17(18)23(27)31-24/h3-12H,1-2H3

InChI key

CHADEQDQBURGHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diacetylfluorescein EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandre Morel et al.
Physiologia plantarum, 152(1), 184-201 (2014-01-28)
Maritime pine somatic embryos (SEs) require a reduction in water availability (high gellan gum concentration in the maturation medium) to reach the cotyledonary stage. This key switch, reported specifically for pine species, is not yet well understood. To facilitate the
Michelle Lane et al.
PloS one, 9(7), e100832-e100832 (2014-07-10)
Paternal health cues are able to program the health of the next generation however the mechanism for this transmission is unknown. Reactive oxygen species (ROS) are increased in many paternal pathologies, some of which program offspring health, and are known
Karen F Chambers et al.
PloS one, 9(11), e111029-e111029 (2014-11-08)
Despite monolayer cultures being widely used for cancer drug development and testing, 2D cultures tend to be hypersensitive to chemotherapy and are relatively poor predictors of whether a drug will provide clinical benefit. Whilst generally more complicated, three dimensional (3D)
Chuanxiong Nie et al.
Biomacromolecules, 16(3), 992-1001 (2015-02-11)
Combining the advantages of the fibrous nanostructure of carbon nanotubes (CNTs) and the bioactivities of heparin/heparin-mimicking polyanions, functional nanofibrous heparin or heparin-mimicking multilayers were constructed on PVDF membrane with highly promoted endothelialization and antithrombogenic activities. Oxidized CNT (oCNT) was first
Ling-Wei Hu et al.
PloS one, 9(5), e97880-e97880 (2014-05-23)
The neurotoxin 6-hydroxydopamine (6-OHDA), which causes transcriptional changes associated with oxidative and proteotoxic stress, has been widely used to generate an experimental model of Parkinson's disease. The food-derived compound luteolin has multi-target actions including antioxidant, anti-inflammatory and neurotrophic activities. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service