Skip to Content
Merck
All Photos(1)

Key Documents

RE-AH8-1

rAra h 8 (Arachis hypogaea allergen 8.0101), peanut

pkg of 250 μg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.41

Pricing and availability is not currently available.

grade

analytical standard

mol wt

Mw 17 kDa

packaging

pkg of 250 μg

manufacturer/tradename

InBio

format

neat

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU111781EHU005781EHU005881
esiRNA cDNA target sequence

TATATGGCTATGGCGGCTTCAACATATCCATCACACCCAACTACAGTGTTTCCAGGCTTATTTTTGTGAGACACATGGGTGGTATCCTGGCAGTGGCCAACATCAGAGGAGGTGGCGAATATGGAGAGACGTGGCATAAAGGTGGTATCTTGGCCAACAAACAAAACTGCTTTGATGACTTTCAGTGTGCTGCTGAGTATCTGATCAAGGAAGGTTACACATCTCCCAAGAGGCTGACTATTAATGGAGGTTCAAATGGAGGCCTCTTAGTGGCTGCTTGTGCAAATCAGAGACCTGACCTCTTTGGTTGTGTTATTGCCCAAGTTGGAGTAATGGACATGCTGAAGTTTCATAAATATACCATCGGCCATGCTTGGACCACTGATTATGGGTGCTCGGACAGCAAACAACACTTTGAATGGCTTGTCAAATACTCTCCATTGCATAATGTGAAGTTACCAGAAGCAGATGACATCCAGTACCCGTCCATGCTGCTCCTCACTGCTGACCATGA

esiRNA cDNA target sequence

GGAACGGAGAGATGGAGTGAACAAAGATTATGAAGAAACCGAGCTTATCAGCACCACAGCTAACTATAGGGCTGTTGGCCCCACTGCTGAGGCGGACAAATCAGCTGCAGAGAAGAGAAGACAGTTGATCCAGGAGTCCAAATTCTTGGGTGGTGACATGGAACACACCCATTTGGTGAAAGGCTTGGATTTTGCTCTGCTTCAAAAGGTACGAGCTGAGATTGCCAGCAAAGAGAAAGAGGAAGAGGAACTGATGGAAAAGCCCCAGAAAGAAACCAAGAAAGATGAGGATCCTGAAAATAAAATTGAATTTAAAACACGTCTGGGCCGCAATGTTTACCGAATGCTTTTTAAGAGCAAAGCATATGAGCGGAATGAGTTGTTCCTGCCGGGCCGCATGGCCTATGTGGTAGACCTGGATGATGAGTATGCTGACACAGATATCCCCA

esiRNA cDNA target sequence

GACCAGCCCTTCATTGATTGTATCAGAGTTGCTCCAGATGAAAAATATGTGGCTGCCAAGATAAGAACTGAAGATTCTGAAGCATCTACCTGTGTAATTATAAAGCTCAGCGATCAGCCCGTAATGGAAGCTTCTTTCCCGAATGTGTCCAGTTTTGAATGGGTAAAGGACGAGGAAGATGAAGATGTTTTATTCTACACCTTCCAGAGGAACCTTCGCTGTCATGACGTATATCGAGCCACTTTTGGTGATAACAAACGTAATGAACGCTTTTACACAGAAAAAGACCCAAGCTACTTTGTTTTCCTTTATCTTACAAAAGACAGTCGTTTCCTCACCATAAATATTATGAACAAGACTACTTCTGAAGTGTGGTTGATAGATGGCCTGAGCCCTTGGGACCCACCAGTACTTATCCAGAAGCGAATACATGGGGT

esiRNA cDNA target sequence

TTCCCATCTGCCCTCTACTGTGATAGCCGCAACCTGCGAAAGGTCCCTGTCATCCCGCCCCGCATCCATTACCTCTATCTCCAGAACAACTTCATCACTGAGCTCCCGGTGGAGTCCTTCCAGAATGCCACAGGCCTGCGATGGATTAACCTGGACAACAACCGAATCCGCAAGATAGACCAGAGGGTGCTGGAGAAACTGCCCGGCCTGGTGTTCCTCTACATGGAGAAGAACCAGTTGGAAGAGGTCCCCTCGGCCCTGCCCCGGAACCTGGAGCAGCTGAGGCTGAGCCAGAACCACATCTCCAGAATCCCGCCTGGTGTCTTCAGCAAGCTGGAGAACCTGCTGCTCCTGGATCTCCAGCACAACAGGCTGAGCGACGGCGTCTTCAAGCCCGACACCTTCCATGGCCTCAAGAACCTCATGCAGCTCAACCT

Gene Information

human ... PREP(5550), PREP(5550)

Gene Information

human ... IK(3550), AC116353.2(3550)

Gene Information

human ... PREPL(9581), PREPL(9581)

Gene Information

human ... PRELP(5549), PRELP(5549)

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

General description

Source: E. coli
Formulation: Preservative-free and carrier-free in phosphate buffered saline, pH 7.4. Steriled filtered.
Concentration: See product insert.
Purification: Purified from E. coli culture by HPLC gel filtration.
Purity > 95 % by silver stained SDS-PAGE.

Disclaimer

Allergens are provided for research and commercial use in vitro:  not for human in vivo or therapeutic use.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service