Skip to Content
Merck
All Photos(1)

Key Documents

PHR1277

Supelco

Nitrilotriacetic acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Nitrilotriacetic acid, N,N-Bis(carboxymethyl)glycine, NTA, Tris(carboxymethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N(CH2COOH)3
CAS Number:
Molecular Weight:
191.14
Beilstein:
1710776
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1463950

API family

edta

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

245 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CN(CC(O)=O)CC(O)=O

InChI

1S/C6H9NO6/c8-4(9)1-7(2-5(10)11)3-6(12)13/h1-3H2,(H,8,9)(H,10,11)(H,12,13)

InChI key

MGFYIUFZLHCRTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitrilotriacetic acid (NTA) is a polyaminopolycarboxylic acid, widely used as a synthetic chelating agent. It forms stable and water-soluble chelates with different metals. It is used as a detergent builder to soften water and for masking metal ions in various industrial processes.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

An excellent chelating agent.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3702 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative gas chromatographic determination of nitrilotriacetic acid in the presence of other carboxylic acids
Larson RA, et al.
Journal of Chromatography A, 111(1), 43-49 (1975)
Determination of nitrilotriacetic acid and ethylenediaminetetraacetic acid in environmental samples as their methyl ester derivatives by gas chromatography-mass spectrometry
Nishikawa Y and Okumura T
Journal of Chromatography A, 690(1), 109-118 (1995)
Determination of the solubility products of nitrilotriacetic acid, ethylenediaminetetraacetic acid and diethylenetriaminepentaacetic acid.
Karhu J, et al.
Analytica Chimica Acta, 380(1), 105-111 (1999)
The determination of part-per-billion levels of citric and nitrilotriacetic acids in tap water and sewage effluents
Aue WA, et al.
Journal of Chromatography A, 72(2), 259-267 (1972)
Asgar Abbas Kazrani et al.
Nucleic acids research, 42(9), 5929-5936 (2014-03-19)
PvuRts1I is a prototype for a larger family of restriction endonucleases that cleave DNA containing 5-hydroxymethylcytosine (5hmC) or 5-glucosylhydroxymethylcytosine (5ghmC), but not 5-methylcytosine (5mC) or cytosine. Here, we report a crystal structure of the enzyme at 2.35 Å resolution. Although

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service