Recommended Products
grade
certified reference material
Quality Level
Agency
suitable for EPA 1633
packaging
pkg of 5 x 8 g
manufacturer/tradename
NIST®
application(s)
PFAS testing
format
matrix material
General description
This Standard Reference Material (SRM) is a frozen fish tissue homogenate that was prepared from fish collected from Lake Michigan, and is intended primarily for use in evaluating analytical methods for the determination of selected trace elements, methylmercury, total mercury, polychlorinated biphenyl (PCB) congeners, chlorinated pesticides, polybrominated diphenyl ether (PBDE) congeners, perfluoroalkyl acids (PFAAs), proximates, a-hexabromocyclododecane (a-HBCD), caloric content, and fatty acids in fish tissue and similar matrices.
A unit of SRM 1947 consists of five bottles, each containing approximately eight grams (wet basis) of frozen tissue homogenate.
For more information, please refer to the COA and SDS.
SRM 1947_cert
SRM 1947_SDS
A unit of SRM 1947 consists of five bottles, each containing approximately eight grams (wet basis) of frozen tissue homogenate.
For more information, please refer to the COA and SDS.
SRM 1947_cert
SRM 1947_SDS
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service