Skip to Content
Merck
All Photos(1)

Key Documents

NIST1888B

Portland cement

NIST® SRM® 1888b

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 4 vials x 5 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 1888b comprises four sealed vials, each containing ~ 5 grams of portland cement ground to a fineness to pass through a 75 µm (No. 200) sieve.


SRM 1888B_cert


SRM 1888B_SDS

Application

SRM1888b is intended primarily for the evaluation or calibration of methods for analysis of cements and materials of similar matrix.

Features and Benefits

  • Accompanied with certificate documenting NIST- certified mass fraction values, and coverage factor for the constituents in portland cement.
  • The reference and information values of constituents are reported as mass fraction.
  • The NIST certificate is provided with expiration certificate, use, storage, handling, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Silicon dioxide (SiO2), Aluminium oxide (Al2O3), Ferric oxide (Fe2O3), Calcium oxide (CaO), Magnesium oxide (MgO), Sulphur Trioxide (SO3), Sodium oxide (Na2O), Potassium oxide (K2O), Titanium dioxide (TiO2), Phosphorus pentoxide (P2O5), Manganese(III) oxide (Mn2O3), Chlorine (Cl), Strontium oxide (SrO)

Reference/Informational value is provided for the following analytes.
Chromium(III) oxide (Cr2O3), Zinc oxide (ZnO), Sulfide Sulfur (S2-),Free Calcium oxide (CaO), Fluoride (F)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service