Skip to Content
Merck
All Photos(1)

Key Documents

K3255

Supelco

N-(β-Ketocaproyl)-DL-homoserine lactone

analytical standard

Synonym(s):

N-(3-Oxohexanoyl)-DL-homoserine lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO4
CAS Number:
Molecular Weight:
213.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

suitability

suitable for manufacturing use

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

−20°C

SMILES string

CCCC(=O)CC(=O)NC1CCOC1=O

InChI

1S/C10H15NO4/c1-2-3-7(12)6-9(13)11-8-4-5-15-10(8)14/h8H,2-6H2,1H3,(H,11,13)

InChI key

YRYOXRMDHALAFL-UHFFFAOYSA-N

Application

N-(β-Ketocaproyl)-DL-homoserine lactone has been used as a reference standard in identifying the quorum sensing signal molecules released by the in wood samples affected by Brenneria salicis bacteria bacteria, using ultra-high performance liquid chromatography coupled with an ion-trap mass spectrometer.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zamia Diaz et al.
PloS one, 4(8), e6820-e6820 (2009-08-29)
Many bacteria produce and respond to the quorum sensing signal autoinducer-2 (AI-2). Escherichia coli and Salmonella typhimurium are among the species with the lsr operon, an operon containing AI-2 transport and processing genes that are up regulated in response to
D Krysciak et al.
Applied and environmental microbiology, 77(15), 5089-5099 (2011-06-07)
Rhizobium sp. strain NGR234 is a unique alphaproteobacterium (order Rhizobiales) that forms nitrogen-fixing nodules with more legumes than any other microsymbiont. Since we have previously described the complete genome sequence of NGR234, we now report on a genome-wide functional analysis
Yunrong Chai et al.
Journal of bacteriology, 191(11), 3706-3711 (2009-03-31)
TraR of Agrobacterium tumefaciens is a LuxR-type quorum-sensing transcription factor that regulates genes required for replication and conjugation of the tumor-inducing (Ti) plasmid. TraR requires its cognate autoinducer N-3-oxooctanoyl-homoserine lactone (OOHL) for resistance of proteolysis in wild-type bacteria and for
Robert J Mitchell et al.
BMB reports, 44(1), 1-10 (2011-01-27)
Inter-cellular communication via diffusible small molecules is a defining character not only of multicellular forms of life but also of single-celled organisms. A large number of bacterial genes are regulated by the change of chemical milieu mediated by the local
Alexandre Crépin et al.
PloS one, 7(4), e35176-e35176 (2012-04-28)
Several small diffusible molecules are involved in bacterial quorum sensing and virulence. The production of autoinducers-1 and -2, quinolone, indole and γ-amino butyrate signaling molecules was investigated in a set of soft-rot bacteria belonging to six Dickeya or Pectobacterium species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service