Skip to Content
Merck
All Photos(1)

Key Documents

ERMEC681M

Polyethylene (high level element content)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

chemicals and industrial polymers

format

matrix material

Analysis Note

For more information please see:
ERMEC681M

Legal Information

ERM is a registered trademark of European Commission

WGK

WGK 3

Flash Point(F)

281.5 °F

Flash Point(C)

138.6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service