Skip to Content
Merck
All Photos(1)

Key Documents

ERMAE642

Chlorine 37Cl (certified for isotope abundance ratio)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

isotopic purity

 37Cl

form

solution

manufacturer/tradename

JRC

format

single component solution

Application


  • Chlorine-37 isotope for tracer studies: The stable isotope of chlorine, 37Cl, is essential in tracer studies to track and understand the dispersal and reaction pathways of chlorine in environmental and biochemical systems, providing precise measurements vital for ecological research (Prieto-Espinoza et al., 2023).

  • Stable chlorine isotope 37Cl for biochemical research: Chlorine-37 is used in biochemical research to study metabolic pathways, helping to elucidate mechanisms of chloride ion transport and chlorine involvement in biochemical reactions, thereby contributing to a deeper understanding of physiological processes (Ojeda et al., 2019).

  • Certified 37Cl isotope abundance for pharma applications: Chlorine-37, certified for its isotope abundance, ensures high precision in pharmaceutical research, particularly in the development of new drugs where isotopic labeling is crucial for tracking and quantifying the fate of chlorinated compounds within biological systems (Wu et al., 2019).

Analysis Note

For more information please see:
ERMAE642

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service