Skip to Content
Merck
All Photos(2)

Key Documents

A7034

Sigma-Aldrich

Bovine Serum Albumin solution

22% in saline, contains azide, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

Quality Level

sterility

aseptically filled

Assay

20.0-24.0% protein basis (biuret)

form

solution

purified by

heat shock fractionation

contains

sodium azide as preservative

does not contain

stabilizer

origin

USA origin

concentration

20.0-24.0% protein (biuret)
22% in saline

UniProt accession no.

foreign activity

BT virus, none detected
VSV virus, none detected

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin is the most abundant plasma protein in humans. Albumin turnover is seen in infants with iron deficiency anemia. Serum albumin is a reliable prognostic indicator for liver disease. Oxidative damage of albumin is associated with advanced liver disease.

Application

Bovine Serum Albumin solution has been used as a component in phosphate-buffered saline (PBS) for antibody dilution.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effects of Gemcitabine and Vinorelbine on inducible nitric oxide synthase (iNOS) and endothelial nitric oxide synthase (eNOS) distribution of MCF-7 breast cancer cells
Zeybek ND, et al.
Acta Histochemica, 113(1), 62-67 (2011)
Zongtang Chu et al.
Artificial cells, nanomedicine, and biotechnology, 48(1), 867-874 (2020-05-29)
Oxygen tension at 50% haemoglobin saturation (P50), which reflects the degree of peripheral oxygen offloading and tissue oxygenation, plays an important role in the diagnosis and treatment of disease, as well as in transfusion research. Blood gas analysers are commonly
Immunohistochemical analysis of small plaque parapsoriasis: involvement of dendritic cells
Zeybek ND, et al.
Acta Histochemica, 110(5), 380-387 (2008)
Neural cell adhesion molecule and neurothelin expression in human ameloblastoma
Er N, et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 59(8), 900-903 (2001)
Molecular Mechanisms of Proteinuria
Molecular and Genetic Basis of Renal Disease, 373-389 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service