Skip to Content
Merck
All Photos(1)

Key Documents

A0420000

Aminocaproic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6-Aminocaproic acid, ε-Aminocaproic acid, 6-Aminohexanoic acid, EACA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)5CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
906872
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

aminocaproic acid

manufacturer/tradename

EDQM

mp

207-209 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NCCCCCC(O)=O

InChI

1S/C6H13NO2/c7-5-3-1-2-4-6(8)9/h1-5,7H2,(H,8,9)

InChI key

SLXKOJJOQWFEFD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Aminocaproic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Lysine analog. Promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis.
Lysine analog. Promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis. Reported to inhibit plasminogen binding to activated platelets. An early report indicated that it inhibits the activation of the first component of the complement system. Binds and inactivates Carboxypeptidase B.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blood-sparing drugs: aprotinin, tranexamic acid, and epsilon-aminocaproic acid.
D Royston
International anesthesiology clinics, 33(1), 155-179 (1995-01-01)
Allergic contact dermatitis due to epsilon-aminocaproic acid: a case report and mini-review of the published work.
Yuriko Yamamoto et al.
The Journal of dermatology, 40(4), 301-303 (2013-02-13)
O N Lucas et al.
Oral surgery, oral medicine, and oral pathology, 51(2), 115-120 (1981-02-01)
On the basis of our review of the literature, it can be concluded that the advantages of E-A.C.A. include (1) safe execution of oral surgical procedures in persons with hemophilia of varying degrees of severity, (2) decrease or total elimination
Are antifibrinolytic agents effective in the treatment of aneurysmal subarachnoid hemorrhage?
Travis R Eastin et al.
Annals of emergency medicine, 64(6), 658-659 (2014-06-24)
Nabaneeta Mukhopadhyay et al.
Physical chemistry chemical physics : PCCP, 17(6), 4293-4310 (2015-01-13)
Blends of polypropylene (PP) and polyamide 6 (PA6) with multiwalled carbon nanotubes (MWNTs) were prepared using different processing strategies in a twin-screw micro-compounder. The effect of MWNTs on the crystallization behaviour of the PP phase and the PA6 phase of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service