Skip to Content
Merck
All Photos(1)

Documents

94882

Supelco

(1S)-(−)-Verbenone

analytical standard

Synonym(s):

(1S,5S)-2-Pinen-4-one, (1S,5S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
1907623
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D −170±12°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.496 (lit.)
n20/D 1.497

bp

227-228 °C (lit.)

density

0.975 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC1=CC(=O)[C@H]2C[C@@H]1C2(C)C

InChI

1S/C10H14O/c1-6-4-9(11)8-5-7(6)10(8,2)3/h4,7-8H,5H2,1-3H3/t7-,8+/m0/s1

InChI key

DCSCXTJOXBUFGB-JGVFFNPUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Monoterpenoid chiral building block.; Isomerization to 2(10)-pinen-4-one; Cyclopentannulation; Oxidative cleavage to pinononic acid

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P.H.J. Carlsen et al.
Acta Chemica Scandinavica. Series B, 38, 501-501 (1984)
G. Ohloff et al.
Helvetica Chimica Acta, 60, 1496-1496 (1977)
K.J. Moriarty et al.
Organometallics, 8, 1512-1512 (1989)
Kevin J Dodds et al.
Journal of economic entomology, 103(6), 2094-2099 (2011-02-12)
Sirex noctilio F. (Hymenoptera: Siricidae) is an invasive woodwasp, currently established in northeastern North America. In other regions of the world, stressed trap trees are used to monitor populations of S. noctilio and to provide inoculation points for the biological
Christina Koutsoudaki et al.
Journal of agricultural and food chemistry, 53(20), 7681-7685 (2005-09-30)
The essential oil and gum of Pistacia lentiscus var. chia, commonly known as the mastic tree, are natural antimicrobial agents that have found extensive uses in medicine in recent years. In this work, the chemical composition of mastic oil and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service