Skip to Content
Merck
All Photos(4)

Key Documents

906700

Sigma-Aldrich

Sodium perchlorate

free-flowing, Redi-Dri, ACS reagent, ≥98.0%

Synonym(s):

Hyperchloric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO4
CAS Number:
Molecular Weight:
122.44
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

grade

ACS reagent

product line

Redi-Dri

Assay

≥98.0%

form

powder

pH

6-8 (25 °C, 5% in water)

mp

468 °C (lit.)

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.02%
Fe: ≤5 ppm
K: ≤0.05%
heavy metals: ≤5 ppm

SMILES string

[Na+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

BAZAXWOYCMUHIX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Redi-Dri is an innovative packaging product line that helps eliminate clumping material by adding extra packaging process steps without any chemical additives, anti-clumping agents, or hydrophobic compounds. Redi-Dri offers a more convenient and safer way to handle the products.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1 - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service