89465
TISAB I solution
Synonym(s):
Total ionic strength adjustment buffer I solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
TISAB, in general, is mixed with samples, as it helps in maintaining the ionic strength and pH constant. It is mainly used to decomplex metal-F complexes. TISAB I can be prepared using NaCl with glacial acetic acid, tri-sodium citrate and NaOH. Visit our Sensor Applications portal to learn more.
Application
TISAB I has been used in determination of fluoride-ion concentration using fluoride ion sensitive electrode. It is an ionic strength buffer, used for the determination of fluoride with ISE in the presence of Ca, Mg, Cl, etc. The pH of the solution is 2-2.5, but can be titrated to pH 5-5.5 by the use of NaOH.
Other Notes
Ionic strength buffer used for the determination of fluoride with ISE in the presence of Ca, Mg, Cl, etc.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Use of a total ionic strength adjustment buffer for electrode determination of fluoride in water supplies.
Analytical chemistry, 40(7), 1169-1171 (1968-06-01)
A study on fluoride sorption by montmorillonite and kaolinite.
Water, Air, Soil Pollut., 141.1-4, 247-261 (2002)
Dependence of Nanotextured Titanium Orthopedic Surfaces on Electrolyte Condition.
Journal of Surface Engineered Materials and Advanced Technology, 6.04, 164-164 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service