Skip to Content
Merck
All Photos(1)

Key Documents

71251

Supelco

Acetic acid

analytical standard

Synonym(s):

Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
E Number:
E260
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.07 (vs air)

vapor pressure

15.2 hPa ( 20.0 °C)
73.3 hPa ( 50.0 °C)

Assay

≥99.8% (GC)

autoignition temp.

800 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

16 %, 92 °F
4 %, 59 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.2% water

evapn. residue

≤0.001%

refractive index

n20/D 1.371 (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

Organoleptic

pungent

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue
F Commodari et al.
NMR in biomedicine, 4(4), 192-200 (1991-08-01)
1H NMR was used to analyze human cerebrospinal fluid (CSF) from a group of neurological disease controls and from a vitamin B12 deficient patient. The spectra were acquired at either 7.06 or 9.40 T at ambient temperature with CSF freeze
Katharine Trenholme et al.
Antimicrobial agents and chemotherapy, 58(7), 3666-3678 (2014-04-16)
Therapies to prevent transmission of malaria parasites to the mosquito vector are a vital part of the global malaria elimination agenda. Primaquine is currently the only drug with such activity; however, its use is limited by side effects. The development
Xianbao Deng et al.
The New phytologist, 201(4), 1469-1483 (2013-11-26)
• Chalcone synthase (CHS) is the key enzyme in the first committed step of the flavonoid biosynthetic pathway and catalyzes the stepwise condensation of 4-coumaroyl-CoA and malonyl-CoA to naringenin chalcone. In plants, CHS is often encoded by a small family
Teagen D Quilichini et al.
The Plant cell, 26(11), 4483-4498 (2014-11-22)
Pollen grains are encased by a multilayered, multifunctional wall. The sporopollenin and pollen coat constituents of the outer pollen wall (exine) are contributed by surrounding sporophytic tapetal cells. Because the biosynthesis and development of the exine occurs in the innermost

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service