Skip to Content
Merck
All Photos(1)

Documents

52517

Supelco

1,1,1,3,3,3-Hexafluoro-2-propanol

for GC derivatization, LiChropur, ≥99.8%

Synonym(s):

HFP, Hexafluoroisopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2CHOH
CAS Number:
Molecular Weight:
168.04
Beilstein:
1841007
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.05

grade

for GC derivatization

Quality Level

Assay

≥99.8% (GC)
≥99.8%

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reagent type: derivatization reagent
reaction type: Esterifications

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.275 (lit.)

bp

59 °C (lit.)

mp

−4 °C (lit.)

density

1.596 g/mL at 25 °C (lit.)

SMILES string

OC(C(F)(F)F)C(F)(F)F

InChI

1S/C3H2F6O/c4-2(5,6)1(10)3(7,8)9/h1,10H

InChI key

BYEAHWXPCBROCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexafluoroisopropanol (HFP) is generally used to denature the native state of proteins, and it also stabilizes the a-helical conformation in unfolded peptides and proteins like ß-lactoglobulin and melittin.

Application

Suitable for the derivatization of glutamic acid, homovanillic, iso-homovanillic and dihydroxyphenyl acetic acids and Υ-aminobutyric acid.

Other Notes

Reagent for hexafluoroisopropyl ester, N-pentafluoropropionyl, and trifluoroacetyl..
Size-exclusion chromatography in 1,1,1,3,3,3-hexafluoro-2-propanol

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1A - STOT RE 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cooperative a-helix formation of ?-lactoglobulin and melittin induced by hexafluoroisopropanol
Hirota, Nami, Yuji Goto, and Kazuko Mizuno.
Protein Science, 6 (2), 416-421 (1997)
Yao Chen et al.
Journal of enzyme inhibition and medicinal chemistry, 33(1), 290-302 (2017-12-28)
Small molecule cholinesterases inhibitor (ChEI) provides an effective therapeutic strategy to treat Alzheimer's disease (AD). Currently, the discovery of new ChEI with multi-target effect is still of great importance. Herein, we report the synthesis, structure-activity relationship study and biological evaluation
Thomas H Mourey et al.
Journal of chromatography. A, 964(1-2), 169-178 (2002-08-30)
1,1,1,3,3,3-Hexafluoroisopropanol is re-examined as an eluent for size-exclusion chromatography (SEC) of polyesters, nylons, and other polar polymers. It is shown that anomalous SEC behavior reported in previous literature can be eliminated by adding 0.01 M tetraethylammonium nitrate to the eluent.
Jie Zhu et al.
Journal of enzyme inhibition and medicinal chemistry, 33(1), 496-506 (2018-02-07)
The cholinergic hypothesis has long been a "polar star" in drug discovery for Alzheimer's disease (AD), resulting in many small molecules and biological drug candidates. Most of the drugs marketed for AD are cholinergic. Herein, we report our efforts in
Sriya Das et al.
ACS applied materials & interfaces, 3(6), 1844-1851 (2011-05-05)
We demonstrate a novel in situ polymerization technique to develop localized polymer coatings on the surface of dispersed pristine graphene sheets. Graphene sheets show great promise as strong, conductive fillers in polymer nanocomposites; however, difficulties in dispersion quality and interfacial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service