Skip to Content
Merck
All Photos(1)

Key Documents

49862

Millipore

Nitrocefin disks

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

−20°C

suitability

Moraxella spp.
Haemophilus spp. (species)
Neisseria spp.
Staphylococcus spp.
anaerobic bacteria

General description

Nitrocefin is a chromogenic cephalosporin, it has a characteristic beta-lactam ring which on oxidation by beta-lactamase, renders nitrocefin from yellow to red in color. It is light-sensitive in nature and is quite expensive. It is used for the rapid detection of beta-lactamase enzymes in isolated colonies of Neisseria gonorrhoeae, Moraxella catarrhalis, Staphylococcus spp., Haemophilus influenzae and anaerobic bacteria.

Application

For the rapid detection of beta-lactamase enzymes in isolated colonies of Neisseria gonorrhoeae, Moraxella catarrhalis, Staphylococcus spp., Haemophilus influenzae and anaerobic bacteria.

Components

50 disks in a light resistant plastic vial (6 mm diameter filter paper disks impregnated with Nitrocefin)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Elizabeth M Adamowicz et al.
The ISME journal, 12(11), 2723-2735 (2018-07-12)
Microbes frequently rely on metabolites excreted by other bacterial species, but little is known about how this cross-feeding influences the effect of antibiotics. We hypothesized that when species rely on each other for essential metabolites, the minimum inhibitory concentration (MIC)
A Piekarowicz et al.
PloS one, 15(10), e0240579-e0240579 (2020-10-16)
All Neisseria gonorrhoeae strains contain multiple copies of integrated filamentous phage genomes with undefined structures. In this study, we sought to characterize the capsid proteins of filamentous N. gonorrhoeae bacteriophage NgoΦ6 and phagemids propagated in different bacteria. The data demonstrate

Articles

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service