Skip to Content
Merck
All Photos(1)

Key Documents

45493

Supelco

Fentin hydroxide

PESTANAL®, analytical standard

Synonym(s):

Triphenyltin hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16OSn
CAS Number:
Molecular Weight:
367.03
Beilstein:
4139186
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O[Sn](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/3C6H5.H2O.Sn/c3*1-2-4-6-5-3-1;;/h3*1-5H;1H2;/q;;;;+1/p-1

InChI key

BFWMWWXRWVJXSE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Fentin hydroxide is a non-systemic fungicide that helps in controlling fungal disease in potatoes, sugar beet, and hops.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David I Shapiro-Ilan et al.
Environmental entomology, 40(1), 59-65 (2011-12-21)
Hypocreales fungi such as Beauveria bassiana (Balsamo) Vuillemin and Metarhizium brunneum Petch can be negatively affected by fungicides thereby reducing their biocontrol potential. In a previous study, we demonstrated enhanced fungicide resistance in B. bassiana through artificial selection. However, it
A Markus et al.
Journal of microencapsulation, 3(1), 39-42 (1986-01-01)
Triphenyltin hydroxide was encapsulated to prolong its fungicidal activity and to decrease its phytotoxicity in peanut fields. The envelopes found to be adequate for this purpose were polyureas and ethyl cellulose, as was demonstrated by biological tests.
G E Walsh et al.
Ecotoxicology and environmental safety, 12(1), 95-100 (1986-08-01)
Effects of water-bourne toxicants on regeneration of arms by the brittle star, Ophioderma brevispina, are described. Regeneration was inhibited by 0.1 micrograms liter-1 bis(tri-n-butyltin)oxide and bis(triphenyltin)oxide. Both substances are known to act upon the nervous system, and it is suggested
Eduardo C Oliveira-Filho et al.
Chemosphere, 81(2), 218-227 (2010-07-03)
Plant molluscicides have been regarded as possible alternatives to the costly and environmentally hazardous molluscicides currently available. This study was undertaken to compare the developmental toxicity of a plant molluscicide (Euphorbia milii latex, LAT) with that of three synthetic molluscicidal
Marcia Sarpa et al.
Regulatory toxicology and pharmacology : RTP, 49(1), 43-52 (2007-07-10)
Triphenyltin-hydroxide (TPTH) is used as agricultural fungicide in Brazil and elsewhere. This study was undertaken to evaluate the developmental toxicity of TPTH in mice. Swiss Webster mice were treated by gavage with TPTH (0, 3.75, 7.5, 15 and 30 mg/kg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service