Skip to Content
Merck
All Photos(1)

Key Documents

34885-M

Sigma-Aldrich

Methanol

≥99.9%, gradient grade, suitable for HPLC

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
15101803
PubChem Substance ID:
NACRES:
NA.05
grade:
gradient grade
technique(s):
HPLC: suitable
bp:
64.7 °C (lit.)
vapor pressure:
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Product Name

Methanol, suitable for HPLC, gradient grade, ≥99.9%

grade

gradient grade

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

technique(s)

HPLC: suitable

impurities

KMnO4 red. matter (as O), in accordance
≤0.0005% free alkali (as NH3)
≤0.0005% non-volatile matter
≤0.001% acetaldehyde
≤0.001% acetone (GC)
≤0.001% formaldehyde
≤0.02% water (Karl Fischer)
≤1 ppb fluorescence (quinine) at 254 nm
≤1 ppb fluorescence (quinine) at 365nm

evapn. residue

<0.0003%

color

APHA: ≤10

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

HPLC-gradient

≤2 mAU at 230 nm
≤5 mAU at 254 nm

absorption

in accordance

UV absorption

λ: 210 nm Amax: 0.50
λ: 220 nm Amax: 0.30
λ: 230 nm Amax: 0.15
λ: 235 nm Amax: 0.10
λ: 240 nm Amax: 0.05
λ: 260 nm Amax: 0.01
λ: 400 nm Amax: 0.01

format

neat

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thermochemical conversion of methanol to C2-C10 hydrocarbons in the presence of shape-selective zeolites has been reported. Its oxidation on Ru-Pt catalyst system by ruthenium ad-atoms has been proposed.

Application

Methanol (MeOH) has been used as an eluent in the HPLC estimation of malondialdehyde in plasma, which is an indicator of oxidative stress.
It may be used in the following studies:
  • Colony forming unit-fibroblast assay of bone marrow mononuclear cells.
  • As solvent for the preparation of extracts of hyphae of Aspergillus for the estimation of gliotoxin by reversed phase-HPLC.
  • Immunofluorescence studies.
  • To compose eluent for the ion-pair reverse-phase HPLC isolation of nucleotides and their decomposition products.

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.
Important notice
  • The article number 34885-6X1L-M will be discontinued. Please order the single bottle 34885-1L-M which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inês Mesquita et al.
Cell reports, 30(12), 4052-4064 (2020-03-27)
Hypoxia-inducible factor-1 alpha (HIF-1α) is considered a global regulator of cellular metabolism and innate immune cell functions. Intracellular pathogens such as Leishmania have been reported to manipulate host cell metabolism. Herein, we demonstrate that myeloid cells from myeloid-restricted HIF-1α-deficient mice
R A Cunha et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 18(6), 1987-1995 (1998-05-15)
ATP analogs substituted in the gamma-phosphorus (ATPgammaS, beta, gamma-imido-ATP, and beta,gamma-methylene-ATP) were used to probe the involvement of P2 receptors in the modulation of synaptic transmission in the hippocampus, because their extracellular catabolism was virtually not detected in CA1 slices.
Ufuk Cobanoglu et al.
Asian Pacific journal of cancer prevention : APJCP, 12(6), 1399-1403 (2011-12-01)
Early diagnosis and prevention is very important for lung cancer patients. Previous studies have emphasized that the level of coenzyme Q10 (CoQ10), present primarily in mitochondria, decreases with age and is low in patients with chronic diseases. Our goal was
Robert E Schwartz et al.
The Journal of clinical investigation, 109(10), 1291-1302 (2002-05-22)
We have derived from normal human, mouse, and rat postnatal bone marrow primitive, multipotent adult progenitor cells (MAPCs) that can differentiate into most mesodermal cells and neuroectodermal cells in vitro and into all embryonic lineages in vivo. Here, we show
Emer P Reeves et al.
Mycopathologia, 158(1), 73-79 (2004-10-19)
Aspergillus fumigatus is a pathogenic fungus capable of causing both allergic lung disease and invasive aspergillosis, a serious, life-threatening condition in neutropenic patients. Aspergilli express an array of mycotoxins and enzymes which may facilitate fungal colonisation of host tissue. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service